Få mere at vide
Benzydamine-d6 Hydrochloride, TRC
Artikelnummer. 30342741
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30342741 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30342741 Leverandør Toronto Research Chemicals Leverandørnr. TRCB20995210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzydamine-d6 Hydrochloride
CAS 1246817-20-8
Molekylær formel C19H18D6ClN3O
Mængde 10 mg
Synonym N,N-(Dimethyl-d6)-3-[[1-(phenylmethyl)-1H-indazol-3-yl]oxy]-1-propanamine Hydrochloride, 1-Benzyl-3-[3-(dimethylamino-d6)propoxy]-1H-indazole Hydrochloride, Afloben-d6, Alcidol-d6, Andolex-d6, Benalgin-d6, Benciflam-d6, Bendaminol-d6, Benzyrin-d6, Difflam-d6, Dorinamin-d6, Enzamin-d6, Imotryl-d6, Riripen-d6, Salyzoron-d6, Saniflor-d6, Tamas-d6, Tantum-d6, Verax-d6
InChI formel InChI=1S/C19H23N3O.ClH/c1-21(2)13-8-14-23-19-17-11-6-7-12-18(17)22(20-19)15-16-9-4-3-5-10-16;/h3-7,9-12H,8,13-15H2,1-2H3;1H/i1D3,2D3;
SMIL [2H]C([2H])([2H])N(C([2H])([2H])[2H])CCCOC1=NN(CC2=CC=CC=C2)C3=CC=CC=C31.Cl
IUPAC navn 3-(1-benzylindazol-3-yl)oxy-N,N-bis(trideuteriomethyl)propan-1-amine;hydrochloride
Molekylvægt (g/mol) 351.9