missing translation for 'onlineSavingsMsg'
Få mere at vide

Benzoyl-d5 Chloride, TRC

Artikelnummer. 30348063
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30348063 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348063 Leverandør Toronto Research Chemicals Leverandørnr. TRCB209553100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Benzoyl-d5 Chloride
CAS 43019-90-5
Molekylær formel C7 2H5 Cl O
Mængde 100 mg
Synonym Benzoyl-2,3,4,5,6-d5 chloride (ACI), Benzoyl-d5 chloride (7CI, 9CI), d5-Benzoyl chloride, Benzoyl Chloride-D5
InChI formel InChI=1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(=O)Cl)c([2H])c1[2H]
IUPAC navn 2,3,4,5,6-pentadeuteriobenzoyl chloride
Molekylvægt (g/mol) 145.6
Formel vægt 145.0343
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.