Få mere at vide
Benzoyl-d5 Chloride, TRC
Artikelnummer. 30348571
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30348571 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348571 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2095531G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzoyl-d5 Chloride
CAS 43019-90-5
Molekylær formel C7 2H5 Cl O
Mængde 1 g
Synonym Benzoyl-2,3,4,5,6-d5 chloride (ACI), Benzoyl-d5 chloride (7CI, 9CI), d5-Benzoyl chloride, Benzoyl Chloride-D5
InChI formel InChI=1S/C7H5ClO/c8-7(9)6-4-2-1-3-5-6/h1-5H/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(=O)Cl)c([2H])c1[2H]
IUPAC navn 2,3,4,5,6-pentadeuteriobenzoyl chloride
Molekylvægt (g/mol) 145.6
Formel vægt 145.0343