Få mere at vide
Benzoxonium Chloride, Mikromol™
Beskrivelse
Benzoxonium Chloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Benzoxonium Chloride |
| CAS | 19379-90-9 |
| Analyt- eller komponentnavne | Benzoxonium Chloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C23 H42 N O2 . Cl |
| Certificeringer/compliance | ISO 17034 |
| Synonym | N-Dodecyl-N,N-bis(2-hydroxyethyl)benzenemethanaminium Chloride (1:1), Ammonium, benzyldodecylbis(2-hydroxyethyl)-, chloride (8CI), Benzenemethanaminium, N-dodecyl-N,N-bis(2-hydroxyethyl)-, chloride (9CI), Benzyldodecylbis(2-hydroxyethyl)ammonium chloride (6CI,7CI), Absonal, Absonal V, Bactofen, Beloran, Benzoxonium chloride, Benzyldodecyldiethanolammonium chloride, Bialcol, Bradophen, Bradophen 100M, Cohortan, D 301, D 301 (antiseptic), Dodecyl(benzyl)diethanolammonium chloride, Dodecylbis(β-hydroxyethyl)benzylammonium chloride, Dodecyldi(β-hydroxyethyl)benzylammonium chloride, Katanol C12, Laurylbis(2-hydroxyethyl)benzylammonium chloride, Lomades, Orofar, Zy 15021 |
| InChI formel | InChI=1S/C23H42NO2.ClH/c1-2-3-4-5-6-7-8-9-10-14-17-24(18-20-25,19-21-26)22-23-15-12-11-13-16-23;/h11-13,15-16,25-26H,2-10,14,17-22H2,1H3;1H/q+1;/p-1 |
| SMIL | [Cl-].CCCCCCCCCCCC[N+](CCO)(CCO)Cc1ccccc1 |
| IUPAC navn | benzyl-dodecyl-bis(2-hydroxyethyl)azanium;chloride |
| Vis mere |