Få mere at vide
Benzotriazole-d4, TRC
Artikelnummer. 30334254
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30334254 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30334254 Leverandør Toronto Research Chemicals Leverandørnr. TRCB20695210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzotriazole-d4
CAS 1185072-03-0
Renhedsgrad noter HPLC
Molekylær formel C6 D4 H N3
Mængde 10 mg
Synonym 1H-Benzotriazole-4,5,6,7-d4, 1H-Benzotriazole D4 (Phenyl D4)
InChI formel InChI=1S/C6H5N3/c1-2-4-6-5(3-1)7-9-8-6/h1-4H,(H,7,8,9)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c2[nH]nnc2c1[2H]
IUPAC navn 4,5,6,7-tetradeuterio-1H-benzotriazole
Molekylvægt (g/mol) 123.149
Formel vægt 123.073
Procent renhed >95