missing translation for 'onlineSavingsMsg'
Learn More

Benzophenone imine, 97%, stabilized, Thermo Scientific Chemicals

Catalog Number p-3735570
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1013-88-3, 123-31-9
C13H11N
181.23
MFCD00001760
SXZIXHOMFPUIRK-UHFFFAOYSA-N
benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
136809
diphenylmethanimine
C1=CC=C(C=C1)C(=N)C2=CC=CC=C2
This item is not returnable. View return policy
1013-88-3, 123-31-9
C13H11N
181.23
MFCD00001760
SXZIXHOMFPUIRK-UHFFFAOYSA-N
benzophenone imine, benzophenoneimine, benzhydrylideneamine, 1,1-diphenylmethanimine, unii-ejj21na7vi, benzhydrylimine, benzophenonimine, diphenylmethanimineimine, 1,1-diphenylmethylimine, benzenemethanimine, .alpha.-phenyl
136809
diphenylmethanimine
C1=CC=C(C=C1)C(=N)C2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 96.0
CAS Max % 100.0
Melting Point -30°C
Color Brown-Yellow to Colorless
Density 1.0800g/mL
Boiling Point 216°C (75.0 mmHg)
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5)2C=NH
Refractive Index 1.6170 to 1.619
Quantity 5 g
Beilstein 07, IV, 1365
Merck Index 15, 1102
Specific Gravity 1.08
Solubility Information Solubility in water: 0.3g/L (25°C). Other solubilities: better soluble in aq. solution at lower ph values,,very soluble in diethyl ether
Stabilizer approx. 0.02% w/w hydroquinone
Viscosity 33,4 mPa.s (20°C)
Formula Weight 181.23
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Benzophenone imine, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzophenone imine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H411-Toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'