Få mere at vide
Benzophenone-2,3,4,5,6-d5, TRC
Artikelnummer. 30343356
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30343356 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343356 Leverandør Toronto Research Chemicals Leverandørnr. TRCB19397010MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzophenone-2,3,4,5,6-d5
CAS 2694-78-2
Molekylær formel C13 D5 H5 O
Mængde 10 mg
Synonym Methanone, phenylphenyl-2,3,4,5,6-d5- (ACI), Benzophenone-2,3,4,5,6-d5 (6CI, 7CI, 8CI), Methanone, phenylphenyl-d5- (9CI), Phenylphenyl-2,3,4,5,6-d5methanone (ACI), 2,3,4,5,6-Pentadeuteriobenzophenone, 2,3,4,5,6-Pentadeuterobenzophenone, Phenyl(phenyl-d5)methanone, Benzophenone-2,3,4,5,6-D5
InChI formel InChI=1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H/i1D,3D,4D,7D,8D
SMIL [2H]c1c([2H])c([2H])c(C(=O)c2ccccc2)c([2H])c1[2H]
IUPAC navn (2,3,4,5,6-pentadeuteriophenyl)-phenylmethanone
Molekylvægt (g/mol) 187.249
Formel vægt 187.105