missing translation for 'onlineSavingsMsg'
Få mere at vide

Benzonitrile-D5, TRC

Artikelnummer. 30343315
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30343315 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30343315 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2046021G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Benzonitrile-D5
CAS 2102-15-0
Molekylær formel C7 D5 N
Mængde 1 g
Synonym Benzonitrile-2,3,4,5,6-d5 (ACI), Benzonitrile-d5 (7CI, 8CI, 9CI), Cyanobenzene-d5, Benzonitrile-D5
InChI formel InChI=1S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5H/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C#N)c([2H])c1[2H]
IUPAC navn 2,3,4,5,6-pentadeuteriobenzonitrile
Molekylvægt (g/mol) 108.152
Formel vægt 108.074
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.