Få mere at vide
Benzonitrile-D5, TRC
Artikelnummer. 30332932
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30332932 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30332932 Leverandør Toronto Research Chemicals Leverandørnr. TRCB204602100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzonitrile-D5
CAS 2102-15-0
Molekylær formel C7 D5 N
Mængde 100 mg
Synonym Benzonitrile-2,3,4,5,6-d5 (ACI), Benzonitrile-d5 (7CI, 8CI, 9CI), Cyanobenzene-d5, Benzonitrile-D5
InChI formel InChI=1S/C7H5N/c8-6-7-4-2-1-3-5-7/h1-5H/i1D,2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C#N)c([2H])c1[2H]
IUPAC navn 2,3,4,5,6-pentadeuteriobenzonitrile
Molekylvægt (g/mol) 108.152
Formel vægt 108.074