Få mere at vide
Benzonatate, TRC
Artikelnummer. 30135162
Skift visning
Mængde:
5 mg
10 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135162 1 g
30007371 5 mg
30012699 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135162 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2045001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 32760-16-0
Molekylær formel (C2H4O)n C12H17NO2
Molekylvægt (g/mol) 193.24
Synonym α-[4-(Butylamino)benzoyl]-ω-methoxy-poly(oxy-1,2-ethanediyl), Benzononantin, Benzononatine, Exangit, KM 65, Tesalon, Tessalin, Tessalon, Tessalon-Ciba, Ventussin;
IUPAC navn 2-methoxyethyl 4-(butylamino)benzoate
SMIL CCCCNC1=CC=C(C(O[(CH2CH2O)nCH3])=O)C=C1
Tekniske data
Kemisk navn eller materiale poly-Benzonatate
Analyt- eller komponentnavne poly-Benzonatate
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C14H21NO3/c1-3-4-9-15-13-7-5-12(6-8-13)14(16)18-11-10-17-2/h5-8,15H,3-4,9-11H2,1-2H3
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 1 g
Formel vægt 603.3619