Få mere at vide
(±)-Benzoin-d10 (diphenyl-d10), CDN
Artikelnummer. 30317145
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30317145 0.25 g -
30310702 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30317145 Leverandør CDN Leverandørnr. CDND58390.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzoin-D10 (diphenyl-D10)
CAS 56830-64-9
Molekylær formel C14 D10 H2 O2
Mængde 0.25 g
Synonym Ethanone, 2-hydroxy-1,2-di(phenyl-d5)- (9 CI), 2-Hydroxy-1,2-di(phenyl-d5)ethanone, Benzoin-d10, Benzoin-D10 (diphenyl-D10)
InChI formel InChI=1 S/C14H12O2/c15-13(11-7-3-1-4-8-11)14(16)12-9-5-2-6-10-12/h1-10,13,15 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(O)C(=O)c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c([2 H])c1[2 H]
IUPAC navn 2-hydroxy-1,2-bis(2,3,4,5,6-pentadeuteriophenyl)ethanone
Molekylvægt (g/mol) 222.3055
Formel vægt 222.1465 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity