Få mere at vide
Benzoic Acid-d5 Ethyl Ester, TRC
Artikelnummer. 30348410
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30348410 100 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348410 Leverandør Toronto Research Chemicals Leverandørnr. TRCB203907100MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzoic Acid-d5 Ethyl Ester
CAS 54354-03-9
Molekylær formel C9H5D5O2
Mængde 100 mg
Synonym Ethyl Benzenecarboxylate-d5, Ethyl Benzoate-d5, NSC 8884-d5
InChI formel InChI=1S/C9H10O2/c1-2-11-9(10)8-6-4-3-5-7-8/h3-7H,2H2,1H3/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(C(=O)OCC)c([2H])c1[2H]
IUPAC navn ethyl 2,3,4,5,6-pentadeuteriobenzoate
Molekylvægt (g/mol) 155.21
Formel vægt 155.0995