Få mere at vide
Benzoestrol, TRC
Artikelnummer. 17980975
Skift visning
Mængde:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17980975 5 mg
17990865 10 mg
17900985 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17980975 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2035505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 85-95-0
Molekylær formel C20 H26 O2
Molekylvægt (g/mol) 298.42
Synonym Phenol, 4,4'-(1,2-diethyl-3-methyl-1,3-propanediyl)bis-, Benzestrol (6CI), Phenol, 4,4'-(1,2-diethyl-3-methyltrimethylene)di- (7CI,8CI), 4,4'-(1,2-Diethyl-3-methyl-1,3-propanediyl)bis[phenol], 3-Ethyl-2,4-bis(p-hydroxyphenyl)hexane, 4,4'-(1,2-Diethyl-3-methyltrimethylene)diphenol, Benzoestrol, Chemestrogen, NSC 408889, Octestrol, Octoestrol, Octofollin, Oktestrol, Phenol, 4,4'-(1,2-diethyl-3-methyl-1,3-propanediyl)bis-, Benzestrol (6CI), Phenol, 4,4'-(1,2-diethyl-3-methyltrimethylene)di- (7CI,8CI), 4,4'-(1,2-Diethyl-3-methyl-1,3-propanediyl)bis[phenol], 3-Ethyl-2,4-bis(p-hydroxyphenyl)hexane, 4,4'-(1,2-Diethyl-3-methyltrimethylene)diphenol, Benzoestrol, Chemestrogen, NSC 408889, Octestrol, Octoestrol, Octofollin, Oktestrol
IUPAC navn 4-[3-ethyl-4-(4-hydroxyphenyl)hexan-2-yl]phenol
SMIL CCC(C(C)c1ccc(O)cc1)C(CC)c2ccc(O)cc2
Tekniske data
Kemisk navn eller materiale Benzoestrol
Analyt- eller komponentnavne Benzoestrol
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C20H26O2/c1-4-19(14(3)15-6-10-17(21)11-7-15)20(5-2)16-8-12-18(22)13-9-16/h6-14,19-22H,4-5H2,1-3H3
Løsningstype Neat
Industry Type Forensics
Mængde 5 mg
Formel vægt 298.1933
Procent renhed >95%