Learn More
Benzoate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™ Product Code.: 12972085
Benzoate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA
Product Code. 12972085 Shop All Fisher Chemical Products
500 mL, Glass Bottle
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Glass Bottle
Product Code. 12972085
Brand: Fisher Chemical A 006 500
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Chemical Identifiers
Molecular Formula C6H5COO-
Specifications
Packaging Glass Bottle
Physical Form Liquid
Concentration 1 g/L
Type Single-component Standard Solution
Solvent or Matrix H2O
Quantity 500 mL
Chemical Name or Material Benzoate
Product Content Correction
Product Title
Benzoate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™ >