Få mere at vide
Benzo[k]fluoranthene-d12, CDN
Artikelnummer. 30319768
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30319768 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319768 Leverandør CDN Leverandørnr. CDND23620.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzo(k)fluoranthene D12
CAS 93952-01-3
Molekylær formel C20 2H12
Mængde 0.01 g
InChI formel InChI=1 S/C20H12/c1-2-6-15-12-19-17-10-4-8-13-7-3-9-16(20(13)17)18(19)11-14(15)5-1/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D
SMIL [2 H]c1c([2 H])c([2 H])c2c([2 H])c3c(c([2 H])c2c1[2 H])c4c([2 H])c([2 H])c([2 H])c5c([2 H])c([2 H])c([2 H])c3c45
IUPAC navn 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuteriobenzo[k]fluoranthene
Molekylvægt (g/mol) 264.38
Formel vægt 264.1692 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity