Få mere at vide
Benzo[ghi]perylene-d12, TRC
Artikelnummer. 30332451
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30332451 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30332451 Leverandør Toronto Research Chemicals Leverandørnr. TRCB2077021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzo(g,h,i)perylene D12
CAS 93951-66-7
Molekylær formel C22 2H12
Mængde 1 mg
Synonym Benzo[ghi]perylene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Benzo[ghi]perylene-d12 (9CI), Perdeuterated benzo[ghi]perylene, Benzo[g,h,i]perylene D12
InChI formel InChI=1S/C22H12/c1-3-13-7-9-15-11-12-16-10-8-14-4-2-6-18-17(5-1)19(13)21(15)22(16)20(14)18/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D
SMIL [2H]c1c([2H])c2c([2H])c([2H])c3c([2H])c([2H])c4c([2H])c([2H])c5c([2H])c([2H])c([2H])c6c(c1[2H])c2c3c4c56
IUPAC navn Benzo[ghi]perylene-1,2,3,4,5,6,7,8,9,10,11,12-d12
Molekylvægt (g/mol) 288.4
Formel vægt 288.1692