Få mere at vide
Benzo[c]phenanthrene-d5, TRC
Artikelnummer. 30331782
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30331782 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30331782 Leverandør Toronto Research Chemicals Leverandørnr. TRCB12190225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzo[c]phenanthrene-d5
CAS 1795011-61-8
Molekylær formel C18 D5 H7
Mængde 25 mg
Synonym 3,4-Benzophenanthrene-d5, Benzo[e]phenanthrene-d5, Tetrahelicene-d5
InChI formel InChI=1S/C18H12/c1-3-7-16-13(5-1)9-11-15-12-10-14-6-2-4-8-17(14)18(15)16/h1-12H/i1D,3D,5D,7D,9D
SMIL [2H]c1cc2ccc3ccccc3c2c4c([2H])c([2H])c([2H])c([2H])c14
IUPAC navn 1,2,3,4,5-pentadeuteriobenzo[c]phenanthrene
Molekylvægt (g/mol) 233.32
Formel vægt 233.125