Få mere at vide
Benzo[b]thiophen-2(3H)-one, TRC
Artikelnummer. 30338182
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30338182 10 g
30342505 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30338182 Leverandør Toronto Research Chemicals Leverandørnr. TRCB20688010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzo[b]thiophen-2(3H)-one
CAS 496-31-1
Molekylær formel C8H6OS
Mængde 10 g
Synonym 2-Oxo-1-thiaindan, 2-Oxo-2,3-dihydrobenzo[b]thiophene, 2-Mercapto-benzeneacetic Acid gamma-(Thio Lactone), Thianaphthen-2-one
InChI formel InChI=1S/C8H6OS/c9-8-5-6-3-1-2-4-7(6)10-8/h1-4H,5H2
SMIL O=C1Cc2ccccc2S1
IUPAC navn 3H-1-benzothiophen-2-one
Molekylvægt (g/mol) 150.2
Formel vægt 150.0139