Få mere at vide
Benzo[b]fluoranthene-d12, CDN
Artikelnummer. 30327457
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30327457 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327457 Leverandør CDN Leverandørnr. CDND23600.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzo(b)fluoranthene D12
CAS 93951-98-5
Molekylær formel C20 2H12
Mængde 0.01 g
Synonym Benz[e]acephenanthrylene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Benzo[b]fluoranthene D12, Benz[e]acephenanthrylene-d12 (9 CI), Benzo[b]fluoranthene-d12
InChI formel InChI=1 S/C20H12/c1-2-7-14-13(6-1)12-19-16-9-4-3-8-15(16)18-11-5-10-17(14)20(18)19/h1-12 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11 D,12 D
SMIL [2 H]c1c([2 H])c([2 H])c2c(c1[2 H])c3c([2 H])c([2 H])c([2 H])c4c5c([2 H])c([2 H])c([2 H])c([2 H])c5c([2 H])c2c34
IUPAC navn Benz[e]acephenanthrylene-1,2,3,4,5,6,7,8,9,10,11,12-d12
Molekylvægt (g/mol) 264.38
Formel vægt 264.1692 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity