Få mere at vide
Benzo[a]pyrene-1,6-quinone, TRC
Artikelnummer. 30137557
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137557 100 mg
30003528 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137557 Leverandør Toronto Research Chemicals Leverandørnr. TRCB205860100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3067-13-8
Molekylær formel C20H10O2
Molekylvægt (g/mol) 282.29
Synonym Benzo[a]pyrene-1,6-dione, 1,6-Dihydrobenzo[a]pyrene-1,6-dione, NSC 30985
IUPAC navn benzo[b]pyrene-1,6-dione
SMIL C1=CC=C2C(=C1)C3=C4C(=CC=C5C4=C(C=C3)C(=O)C=C5)C2=O
Tekniske data
Kemisk navn eller materiale Benzo[a]pyrene-1,6-quinone
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C20H10O2/c21-17-10-6-11-5-7-16-19-13(8-9-15(17)18(11)19)12-3-1-2-4-14(12)20(16)22/h1-10H
Formel vægt 282.068
Procent renhed >95%