Få mere at vide
Benzimidazole-4,5,6,7-d4, CDN
Artikelnummer. 30319181
Skift visning
Mængde:
0.05 g
0.1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319181 0.1 g
30311390 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319181 Leverandør CDN Leverandørnr. CDND40790.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzimidazole-4,5,6,7-D4
CAS 64531-29-9
Molekylær formel C7 D4 H2 N2
Mængde 0.1 g
Synonym 1 H-Benzimidazole-4,5,6,7-d4 (9 CI), Benzimidazole-4,5,6,7-d4, Benzimidazole-d4, Benzimidazole-4,5,6,7-D4
InChI formel InChI=1 S/C7H6N2/c1-2-4-7-6(3-1)8-5-9-7/h1-5 H,(H,8,9)/i1D,2 D,3 D,4 D
SMIL [2 H]c1c([2 H])c([2 H])c2[nH]cnc2c1[2 H]
IUPAC navn 4,5,6,7-tetradeuterio-1 H-benzimidazole
Molekylvægt (g/mol) 122.1606
Formel vægt 122.0782 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity