Learn More
Benzhydryl isothiocyanate, 97%, Thermo Scientific Chemicals
Product Code. 11364126
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11364126 1 g 1g
11374126 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 11364126 Supplier Thermo Scientific Alfa Aesar Supplier No. L19249.03
 Restricted Item
På lager
Add to basket
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
CAS: 3550-21-8 | Molecular Formula: C14H11NS | Molecular Weight: 225.31 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3550-21-8
Molecular Formula C14H11NS
Molecular Weight (g/mol) 225.31
MDL Number MFCD00046815
InChI Key WDOSFTZMBFYTED-UHFFFAOYSA-N
Synonym benzhydryl isothiocyanate, isothiocyanatodiphenylmethane, benzhydrylisothiocyanate, isothiocyanato phenyl methyl benzene, diphenylmethylisothiocyanate, benzene, 1,1'-isothiocyanatomethylene bis, diphenylmethanisothiocyanate, acmc-20alq6, benzhydryl-isothiocyanate, benzhydryl isothio-cyanate
PubChem CID 520742
IUPAC Name [isothiocyanato(phenyl)methyl]benzene
SMILES S=C=NC(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 58°C to 60°C
Boiling Point 160°C to 165°C (4 mmHg)
Quantity 1 g
UN Number UN2811
Beilstein 2210613
Sensitivity Moisture sensitive
Formula Weight 225.31
Percent Purity 97%
Chemical Name or Material Benzhydryl isothiocyanate