Få mere at vide
Benzhydrol-d10, TRC
Artikelnummer. 30332544
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30332544 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30332544 Leverandør Toronto Research Chemicals Leverandørnr. TRCB19380210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzhydrol-d10
CAS 74563-01-2
Molekylær formel C13H2D10O
Mængde 10 mg
Synonym alpha-Phenylbenzenemethanol-d10, Benzhydryl Alcohol-d10, Benzohydrol-d10, Diphenylcarbinol-d10, Diphenylmethanol, -d10 Diphenylmethyl Alcohol-d10, Hydroxydiphenylmethane-d10, NSC 32150-d10, alpha-Phenylbenzenemethanol-d10, alpha-Phenylbenzyl Alcohol-d10
InChI formel InChI=1S/C13H12O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13-14H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D
SMIL [2H]c1c([2H])c([2H])c(C(O)c2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]
IUPAC navn bis(phenyl-d5)methanol
Molekylvægt (g/mol) 194.3
Formel vægt 194.1516