Få mere at vide
Benzene-13C6, TRC
Artikelnummer. 30135048
Skift visning
Mængde:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135048 500 mg
30132527 100 mg
30003640 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135048 Leverandør Toronto Research Chemicals Leverandørnr. TRCB185283500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 32488-44-1
Molekylær formel 13C6 H6
Molekylvægt (g/mol) 84.07
Synonym Benzene-1,2,3,4,5,6-13C6, Benzene-13C6 (7CI,9CI), 13C6-Benzene, [U-13C]Benzene, Benzene 13C6
IUPAC navn (1,2,3,4,5,6-^{13}C_{6})cyclohexatriene
SMIL [13cH]1[13cH][13cH][13cH][13cH][13cH]1
Tekniske data
Kemisk navn eller materiale Benzene 13C6
Mængde 500 mg
InChI formel InChI=1S/C6H6/c1-2-4-6-5-3-1/h1-6H/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vægt 84.0671