Få mere at vide
Benzene-1,2,4,5-d4, CDN
Artikelnummer. 30319857
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30319857 1 g -
30310734 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319857 Leverandør CDN Leverandørnr. CDND04871G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzene-1,2,4,5-D4
CAS 14919-17-6
Molekylær formel C6 D4 H2
Mængde 1 g
Synonym Benzene-1,2,4,5-d4 (7 CI, 8 CI, 9 CI, ACI), 1,2,4,5-Tetradeuterobenzene, p-Benzene-d4, Benzene-1,2,4,5-D4
InChI formel InChI=1 S/C6H6/c1-2-4-6-5-3-1/h1-6 H/i1D,2 D,5 D,6 D
SMIL [2 H]c1cc([2 H])c([2 H])cc1[2 H]
IUPAC navn 1,2,4,5-tetradeuteriobenzene
Molekylvægt (g/mol) 82.1365
Formel vægt 82.0721 g/mol
Procent renhed 97 atom % D, min 98% Chemical Purity