Få mere at vide
(Benzamido)oxy Acetic Acid-d2, TRC
Artikelnummer. 30330736
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30330736 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30330736 Leverandør Toronto Research Chemicals Leverandørnr. TRCB18510250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale (Benzamido)oxy Acetic Acid-d2
CAS 1346599-67-4
Molekylær formel C9H7D2NO4
Mængde 50 mg
Synonym 2-[(Benzoylamino)oxy]-acetic Acid-d2, (Benzamidooxy)acetic Acid-d2, Benzadox-d2, N-Benzoyl-O-(carboxymethyl)-hydroxylamine-d2, NSC 75601-d2, S 6173 -d2
InChI formel InChI=1S/C9H9NO4/c11-8(12)6-14-10-9(13)7-4-2-1-3-5-7/h1-5H,6H2,(H,10,13)(H,11,12)/i6D2
SMIL [2H]C([2H])(ONC(=O)c1ccccc1)C(=O)O
IUPAC navn 2-benzamidooxy-2,2-dideuterioacetic acid
Molekylvægt (g/mol) 197.18
Formel vægt 197.0657