Learn More
Benzamide, 99%, Thermo Scientific Chemicals
Catalog Number p-62758
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
55-21-0
C7H7NO
121.14
MFCD00007968
KXDAEFPNCMNJSK-UHFFFAOYSA-N
benzoic acid amide, benzoylamide, phenylcarboxyamide, phenylcarboxamide, benzenecarboxamide, amid kyseliny benzoove, amid kyseliny benzoove czech, phenyl carboxyamide, ccris 4594, benzoic acid,amide
2331
CHEBI:28179
benzamide
C1=CC=C(C=C1)C(=O)N
55-21-0
C7H7NO
121.14
MFCD00007968
KXDAEFPNCMNJSK-UHFFFAOYSA-N
benzoic acid amide, benzoylamide, phenylcarboxyamide, phenylcarboxamide, benzenecarboxamide, amid kyseliny benzoove, amid kyseliny benzoove czech, phenyl carboxyamide, ccris 4594, benzoic acid,amide
2331
CHEBI:28179
benzamide
C1=CC=C(C=C1)C(=O)N
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 125.0°C to 129.0°C
Color White
Boiling Point 228.0°C
Flash Point 180°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula C6H5CONH2
Quantity 100 g
Beilstein 09, 195
Merck Index 15, 1062
Solubility Information Solubility in water: 1g/74mL (20°C). Other solubilities: soluble in hot water,soluble in hot benzene,1g/6mL alcohol- 1g/3.3mL pyridine,slightly soluble in ether
Formula Weight 121.14
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Benzamide, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Benzamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.