Få mere at vide
Benzamide-2,3,4,5,6-d5, CDN
Artikelnummer. 30321979
Skift visning
Mængde:
0.25 g
0.5 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30321979 0.25 g -
30310121 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30321979 Leverandør CDN Leverandørnr. CDND60300.25G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzamide-D5
CAS 1235489-47-0
Molekylær formel C7 D5 H2 N O
Mængde 0.25 g
Synonym Benzamide-2,3,4,5,6-d5 (ACI), Benzamide-d5, Benzene-2,3,4,5,6-d5-carboxamide, Benzoic acid amide-d5, Benzamide-D5
InChI formel InChI=1 S/C7H7NO/c8-7(9)6-4-2-1-3-5-6/h1-5 H,(H2,8,9)/i1D,2 D,3 D,4 D,5 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)N)c([2 H])c1[2 H]
IUPAC navn 2,3,4,5,6-pentadeuteriobenzamide
Molekylvægt (g/mol) 126.1674
Formel vægt 126.0841 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity