Få mere at vide
Benzaldehyde-alpha-d1, TRC
Artikelnummer. 30335281
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30335281 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30335281 Leverandør Toronto Research Chemicals Leverandørnr. TRCB11974150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzaldehyde-alpha-d1
CAS 3592-47-0
Molekylær formel C7 D H5 O
Mængde 50 mg
Synonym Benzaldehyde-formyl-d (8CI, 9CI, ACI), Benzaldehyde-α-d (7CI), 1-d1-Benzaldehyde, Benzaldehyde-1-d, Benzaldehyde-carbonyl-d, Benzaldehyde-α-d1, Deuterio(phenyl)methanone, α-Deuteriobenzaldehyde, Benzaldehyde-α-D
InChI formel InChI=1S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6H/i6D
SMIL [2H]C(=O)c1ccccc1
IUPAC navn deuterio(phenyl)methanone
Molekylvægt (g/mol) 107.1281
Formel vægt 107.0481