Få mere at vide
Benzaldehyde-alpha-d1, CDN
Artikelnummer. 30312064
Skift visning
Mængde:
0.5 g
1 g
Pakningsstørrelse:
0.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30312064 1 g -
30328255 0.5 g 0.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312064 Leverandør CDN Leverandørnr. CDND01671G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benzaldehyde-alpha-d1
CAS 3592-47-0
Molekylær formel C7 D H5 O
Mængde 1 g
Synonym Benzaldehyde-formyl-d (8 CI, 9 CI, ACI), Benzaldehyde-α-d (7 CI), 1-d1-Benzaldehyde, Benzaldehyde-1-d, Benzaldehyde-carbonyl-d, Benzaldehyde-α-d1, Deuterio(phenyl)methanone, α-Deuteriobenzaldehyde, Benzaldehyde-α-D
InChI formel InChI=1 S/C7H6O/c8-6-7-4-2-1-3-5-7/h1-6 H/i6D
SMIL [2 H]C(=O)c1ccccc1
IUPAC navn deuterio(phenyl)methanone
Molekylvægt (g/mol) 107.1281
Formel vægt 107.0481 g/mol
Procent renhed 98 atom % D, min 96% Chemical Purity