Få mere at vide
Benz[a]anthracene-7-methanol, TRC
Artikelnummer. 30335088
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30335088 500 mg
30347812 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30335088 Leverandør Toronto Research Chemicals Leverandørnr. TRCB183605500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benz[a]anthracene-7-methanol
CAS 16110-13-7
Molekylær formel C19H14O
Mængde 500 mg
Synonym 7-(Hydroxymethyl)benz[a]anthracene
InChI formel InChI=1S/C19H14O/c20-12-19-16-8-4-2-6-14(16)11-18-15-7-3-1-5-13(15)9-10-17(18)19/h1-11,20H,12H2
SMIL OCc1c2ccccc2cc3c1ccc4ccccc34
IUPAC navn benzo[a]anthracen-7-ylmethanol
Molekylvægt (g/mol) 258.31
Formel vægt 258.1045