Få mere at vide
Bentazon-d7 (iso-propyl-d7), CDN
Artikelnummer. 30323194
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323194 0.01 g
30311055 0.005 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323194 Leverandør CDN Leverandørnr. CDND70980.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bentazon-d7
CAS 131842-77-8
Molekylær formel C10 D7 H5 N2 O3 S
Mængde 0.01 g
Synonym Bentazone D7 (isopropyl D7), 1 H-2,1,3-Benzothiadiazin-4(3 H)-one, 3-[1-(methyl-d3)ethyl-1,2,2,2-d4]-, 2,2-dioxide (9 CI), 3-Isopropyl-d7-1 H-2,1,3-benzothiadiazin-4(3 H)-one 2,2-dioxide
InChI formel InChI=1 S/C10H12N2O3S/c1-7(2)12-10(13)8-5-3-4-6-9(8)11-16(12,14)15/h3-7,11 H,1-2H3/i1D3,2D3,7 D
SMIL [2 H]C([2 H])([2 H])C([2 H])(N1C(=O)c2ccccc2NS1(=O)=O)C([2 H])([2 H])[2 H]
IUPAC navn 3-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-2,2-dioxo-1 H-2λ^{6},1,3-benzothiadiazin-4-one
Molekylvægt (g/mol) 247.322
Formel vægt 247.101 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity