Beskrivelse
Honeywell Fluka™ offers an expanding range of high quality inorganic certified reference materials and organic reference materials.
- Produced according to ISO 9001
- Suitable for a wide range of industries and applications
- Detailed Certificate of Analysis included with every delivery
- Expert Technical Support
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bentazon-d7 |
| Emballage | Glass Bottle |
| CAS | 131842-77-8 |
| Farve | White |
| Smeltepunkt | 131°C to 134°C |
| Synonym | 3-Isopropyl-d7-1H-2,1,3-benzothiadiazin-4(3H)-one |
| Industry Type | Environmental Analysis |
| Molekylvægt (g/mol) | 247.32 |
| Mængde | 10 mg |
| Grad | Analytical Standard |