missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Bensultap 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Kemisk navn eller materiale | Bensultap |
| CAS | 17606-31-4 |
| Analyt- eller komponentnavne | Bensultap |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C17 H21 N O4 S4 |
| Koncentration | 100 μg/mL |
| Certificeringer/compliance | ISO 17034 |
| InChI formel | InChI=1S/C17H21NO4S4/c1-18(2)15(13-23-25(19,20)16-9-5-3-6-10-16)14-24-26(21,22)17-11-7-4-8-12-17/h3-12,15H,13-14H2,1-2H3 |
| SMIL | CN(C)C(CSS(=O)(=O)c1ccccc1)CSS(=O)(=O)c2ccccc2 |
| IUPAC navn | 1,3-bis(benzenesulfonylsulfanyl)-N,N-dimethylpropan-2-amine |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?