Få mere at vide
Benproperine Phosphate, TRC
Artikelnummer. 30347488
Skift visning
Mængde:
5 g
Artikelnummer. Mængde
30347488 5 g
1 options
Artikelnummer. 30347488 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1615005G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benproperine Phosphate
CAS 19428-14-9
Molekylær formel C21 H27 N O . H3 O4 P
Mængde 5 g
Synonym Piperidine, 1-[1-methyl-2-[2-(phenylmethyl)phenoxy]ethyl]-, phosphate (1:1), Piperidine, 1-[1-methyl-2-[(α-phenyl-o-tolyl)oxy]ethyl]-, phosphate (1:1) (8CI), 1-(2-Benzylphenoxy)-2-piperidinopropane phosphate, 1-[2-(2-Benzylphenoxy)-1-methylethyl]piperidine monophosphate, ASA 158-5, Benproperine phosphate, Blascorid, Pirexyl
InChI formel InChI=1S/C21H27NO.H3O4P/c1-18(22-14-8-3-9-15-22)17-23-21-13-7-6-12-20(21)16-19-10-4-2-5-11-19;1-5(2,3)4/h2,4-7,10-13,18H,3,8-9,14-17H2,1H3;(H3,1,2,3,4)
SMIL CC(COc1ccccc1Cc2ccccc2)N3CCCCC3.OP(=O)(O)O
IUPAC navn 1-[1-(2-benzylphenoxy)propan-2-yl]piperidine;phosphoric acid
Molekylvægt (g/mol) 407.44
Formel vægt 407.1862