Få mere at vide
Benoxaprofen-13C,d3, TRC
Artikelnummer. 30341507
Skift visning
Mængde:
25 mg
Artikelnummer. Mængde
30341507 25 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341507 Leverandør Toronto Research Chemicals Leverandørnr. TRCB16530225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benoxaprofen-13C,d3
CAS 1329840-53-0
Molekylær formel C1513CH9D3ClNO3
Mængde 25 mg
Synonym 2-(4-Chlorophenyl)-alpha-(methyl-13C,d3)-5-benzoxazoleacetic Acid, (RS)-Benoxaprofen-13C,d3, (+/-)-Benoxaprofen-13C,d3, 2-[2-(4-Chlorophenyl)benzoxazol-5-yl]propanoic Acid-13C,d3, Compound 90459-13C,d3, Coxigon-13C,d3, LRCL 3794-13C,d3, Lilly 90459-13C,d3, NSC 299582-13C,d3, Opren-13C,d3, Oraflex-13C,d3, Uniprofen-13C,d3, dl-Benoxaprofen-13C,d3
InChI formel InChI=1S/C16H12ClNO3/c1-9(16(19)20)11-4-7-14-13(8-11)18-15(21-14)10-2-5-12(17)6-3-10/h2-9H,1H3,(H,19,20)/i1+1D3
SMIL [2H][13C]([2H])([2H])C(C(=O)O)c1ccc2oc(nc2c1)c3ccc(Cl)cc3
IUPAC navn 2-[2-(4-chlorophenyl)-1,3-benzoxazol-5-yl]-3,3,3-trideuterio(3^{13}C)propanoic acid
Molekylvægt (g/mol) 305.74
Formel vægt 305.0728