Få mere at vide
Benomyl-d4 (benzimidazole-4,5,6,7-d4), TRC
Artikelnummer. 30347526
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30347526 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30347526 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1613815MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benomyl-d4 (benzimidazole-4,5,6,7-d4)
CAS 1398065-98-9
Molekylær formel C14 D4 H14 N4 O3
Mængde 5 mg
Synonym Benomyl-d4, Benomyl-d4 (benzimidazole-4,5,6,7-d4), Benomyl-D4 (benzimidazole-4,5,6,7-D4), Methyl 1-(butylcarbamoyl)-2-benzimidazolyl-4,5,6,7-d4-carbamate, [1-[(Butylamino)carbonyl]-1H-benzimidazol-2-yl-4,5,6,7-d4]carbamic acid methyl ester
InChI formel InChI=1S/C14H18N4O3/c1-3-4-9-15-13(19)18-11-8-6-5-7-10(11)16-12(18)17-14(20)21-2/h5-8H,3-4,9H2,1-2H3,(H,15,19)(H,16,17,20)/i5D,6D,7D,8D
SMIL [2H]c1c([2H])c([2H])c2c(nc(NC(=O)OC)n2C(=O)NCCCC)c1[2H]
IUPAC navn methyl N-[1-(butylcarbamoyl)-4,5,6,7-tetradeuteriobenzimidazol-2-yl]carbamate
Molekylvægt (g/mol) 294.342
Formel vægt 294.163