Få mere at vide
Bendiocarb, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Bendiocarb |
| CAS | 22781-23-3 |
| Molekylær formel | C11 H13 N O4 |
| Mængde | 1 g |
| Synonym | 1,3-Benzodioxol-4-ol, 2,2-dimethyl-, methylcarbamate (9CI), Carbamic acid, methyl-, 2,3-(isopropylidenedioxy)phenyl ester (8CI), Phenol, 2,3-(isopropylidenedioxy)-, methylcarbamate (8CI), 2,2-Dimethyl-1,3-benzodioxol-4-yl N-methylcarbamate, 2,2-Dimethyl-1,3-benzodioxol-4-yl methylcarbamate, Bencarbate, Bendiocarb, Dycarb, Ficam, Ficam 80W, Ficam W, Fisons NC 6897, Garvox, Multamat, NC 6897, Niomil, Seedox, Seedox (insecticide), Seedox 80W, Tatoo, Tattoo, Turcam |
| InChI formel | InChI=1S/C11H13NO4/c1-11(2)15-8-6-4-5-7(9(8)16-11)14-10(13)12-3/h4-6H,1-3H3,(H,12,13) |
| SMIL | CNC(=O)Oc1cccc2OC(C)(C)Oc12 |
| IUPAC navn | (2,2-dimethyl-1,3-benzodioxol-4-yl) N-methylcarbamate |
| Molekylvægt (g/mol) | 223.23 |
| Formel vægt | 223.0845 |