missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Bendamustine Ethyl Ester, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Bendamustine Ethyl Ester |
| CAS | 87475-54-5 |
| Molekylær formel | C18 H25 Cl2 N3 O2 |
| Mængde | 250 mg |
| Synonym | 1H-Benzimidazole-2-butanoic acid, 5-[bis(2-chloroethyl)amino]-1-methyl-, ethyl ester, Bendamustine ethyl ester, Ethyl 5-[bis(2-chloroethyl)amino]-1-methyl-1H-benzimidazole-2-butanoate, Bendamustine Ethyl Ester |
| InChI formel | InChI=1S/C18H25Cl2N3O2/c1-3-25-18(24)6-4-5-17-21-15-13-14(7-8-16(15)22(17)2)23(11-9-19)12-10-20/h7-8,13H,3-6,9-12H2,1-2H3 |
| SMIL | CCOC(=O)CCCc1nc2cc(ccc2n1C)N(CCCl)CCCl |
| IUPAC navn | ethyl 4-[5-[bis(2-chloroethyl)amino]-1-methylbenzimidazol-2-yl]butanoate |
| Molekylvægt (g/mol) | 386.32 |
| Formel vægt | 385.1324 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?