Få mere at vide
Kern Sohn Bench Scale GAB
Artikelnummer. p-7226297
Encompass_Preferred
Volumen:
12 kg
15 kg, 30 kg
3 kg, 6 kg
30 kg
6 kg
6 kg, 15 kg
Pakningsstørrelse:
Hver
Artikelnummer. 17381653
Brand: Kern Sohn GAB 12K0.1N
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
This item requires a price update prior to being ordered.
Request a Price for this item here.
, 02 950 59 478
Compact bench scale that is practical for small spaces.
  • Compact size, practical for small spaces
  • High mobility: thanks to rechargeable battery operation (optional), compact, lightweight construction, it is suitable for the use in several locations (production, warehouse, dispatch department etc.)
  • Weighing with tolerance range (check-weighing): a visual and audible signal helps with portioning, dispensing or grading
  • Totalising of weights
  • Protective working cover included with delivery
Tekniske data
Volumen 12 kg
Læsbarhed 0.1 g
Model- Compact
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.