Få mere at vide
Kern Sohn Bench Scale FKB
Artikelnummer. p-7226295
Encompass_Preferred
Volumen:
6 kg
8 kg
15 kg
16 kg
30 kg
36 kg
65 kg
Læsbarhed:
0.02 g
0.05 g
0.1 g
0.2 g
0.5 g
1 g
Artikelnummer. 17391633
Brand: Kern Sohn FKB 15K0.5
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
High-resolution bench scale with large weighing range and robust stainless steel weighing plate.
  • Thanks to the high resolution it is ideal for high-precision weighing in the industrial field
  • Housing with robust aluminum diecast housing trough, maintains the stability, protects the weighing technology elements and is robust enough to cope with everyday use
  • Particularly practical: thanks to the large weighing ranges and compact dimensions, you can accurately weigh heavy loads in the most restricted of spaces. Useful for determining very small weight differences, such as, for example, gas wastage, abrasion of mechanical parts, rock samples, minerals, druses, silver etc.
  • PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
  • Freely programmable weighing unit, e.g. display direct in special units such as length of wire g/m, surface weight g/m2, or else
  • Protective working cover included with delivery
Tekniske data
Volumen 15 kg
Læsbarhed 0.5 g
Materiale Stainless Steel
Korrektion af produktindhold
Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.