Learn More
Kern Sohn Bench Scale FKB
High-resolution bench scale with large weighing range and robust stainless steel weighing plate.
Encompass_Preferred
POA - 1972.62 DKK
Specifications
Capacity 15 kg
Material Stainless Steel
View More Specs
Products 2
Product Code Brand Capacity Readability Price Quantity & Availability  
Product Code Brand Capacity Readability Price Quantity & Availability  
17301643
Encompass_Preferred
View Documents
Kern Sohn
FKB 15K0.5A
15 kg 0.5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
17311643
Encompass_Preferred
View Documents
Kern Sohn
FKB 15K1A
15 kg 1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Thanks to the high resolution it is ideal for high-precision weighing in the industrial field
  • Housing with robust aluminum diecast housing trough, maintains the stability, protects the weighing technology elements and is robust enough to cope with everyday use
  • Particularly practical: thanks to the large weighing ranges and compact dimensions, you can accurately weigh heavy loads in the most restricted of spaces. Useful for determining very small weight differences, such as, for example, gas wastage, abrasion of mechanical parts, rock samples, minerals, druses, silver etc.
  • PRE-TARE function for manual subtraction of a known container weight, useful for checking fill-levels
  • Freely programmable weighing unit, e.g. display direct in special units such as length of wire g/m, surface weight g/m2, or else
  • Protective working cover included with delivery
Specifications
Specifications
15 kg
Stainless Steel
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Kern Sohn Bench Scale FKB