Få mere at vide
Benazeprilat-d5, TRC
Artikelnummer. 30134519
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134519 1 mg
17939439 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30134519 Leverandør Toronto Research Chemicals Leverandørnr. TRCB1197621MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1279033-05-4
Molekylær formel C22H19D5N2O5
Molekylvægt (g/mol) 401.47
Synonym (3S)-3-[[(1S)-1-Carboxy-3-phenylpropyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic Acid-d5;
IUPAC navn (2S)-2-[[(3S)-1-(carboxymethyl)-2-oxo-4,5-dihydro-3H-1-benzazepin-3-yl]amino]-4-(2,3,4,5,6-pentadeuteriophenyl)butanoic acid
SMIL O=C1[C@@H](N[C@H](C(O)=O)CCC2=C([2H])C([2H])=C([2H])C([2H])=C2[2H])CCC3=C(C=CC=C3)N1CC(O)=O
Tekniske data
Kemisk navn eller materiale Benazeprilat-d5
Mængde 1 mg
InChI formel InChI=1S/C22H24N2O5/c25-20(26)14-24-19-9-5-4-8-16(19)11-13-17(21(24)27)23-18(22(28)29)12-10-15-6-2-1-3-7-15/h1-9,17-18,23H,10-14H2,(H,25,26)(H,28,29)/t17-,18-/m0/s1/i1D,2D,3D,6D,7D
Formel vægt 401.1999