Få mere at vide
Benazepril-d5 Hydrochloride, TRC
Artikelnummer. 30348166
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30348166 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30348166 Leverandør Toronto Research Chemicals Leverandørnr. TRCB11975310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Benazepril-d5 Hydrochloride
CAS 1279026-26-4
Molekylær formel C24H24D5ClN2O5
Mængde 10 mg
Synonym (3S)-3-[[(1S)-1-(Ethoxycarbonyl)-3-phenyl-d5-propyl]amino]-2,3,4,5-tetrahydro-2-oxo-1H-1-benzazepine-1-acetic Acid Hydrochloride, Lotensin-d5
InChI formel InChI=1S/C24H28N2O5.ClH/c1-2-31-24(30)20(14-12-17-8-4-3-5-9-17)25-19-15-13-18-10-6-7-11-21(18)26(23(19)29)16-22(27)28;/h3-11,19-20,25H,2,12-16H2,1H3,(H,27,28);1H/t19-,20-;/m0./s1/i3D,4D,5D,8D,9D;
SMIL O=C(OCC)[C@@H](N[C@@H]1C(N(CC(O)=O)C(C=CC=C2)=C2CC1)=O)CCC3=C([2H])C([2H])=C([2H])C([2H])=C3[2H].Cl
IUPAC navn 2-[(3S)-3-[[(2S)-1-ethoxy-1-oxo-4-(2,3,4,5,6-pentadeuteriophenyl)butan-2-yl]amino]-2-oxo-4,5-dihydro-3H-1-benzazepin-1-yl]acetic acid;hydrochloride
Molekylvægt (g/mol) 465.98