Få mere at vide
Benactyzine N-Oxide, Mikromol™
Beskrivelse
Benactyzine N-Oxide
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Benactyzine N-Oxide |
| CAS | 42771-70-0 |
| Analyt- eller komponentnavne | Benactyzine N-Oxide |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C20 H25 N O4 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Benactyzine N-Oxide, Benzeneacetic acid, alpha-hydroxy-alpha-phenyl-, 2-(diethyloxidoamino)ethyl ester, Benzeneacetic acid, alpha-hydroxy-alpha-phenyl-, 2-(diethylamino)ethyl ester, N-oxide |
| InChI formel | InChI=1S/C20H25NO4/c1-3-21(24,4-2)15-16-25-19(22)20(23,17-11-7-5-8-12-17)18-13-9-6-10-14-18/h5-14,23H,3-4,15-16H2,1-2H3 |
| SMIL | CC[N+]([O-])(CC)CCOC(=O)C(O)(c1ccccc1)c2ccccc2 |
| IUPAC navn | N,N-diethyl-2-(2-hydroxy-2,2-diphenylacetyl)oxyethanamine oxide |
| Vis mere |