Få mere at vide
Bempedoic Acid-d5, TRC
Artikelnummer. 30341942
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30341942 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30341942 Leverandør Toronto Research Chemicals Leverandørnr. TRCB11970150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Bempedoic Acid-d5
CAS 2408131-71-3
Molekylær formel C19 H31 D5 O5
Mængde 50 mg
Synonym 8-Hydroxy-2,2,14,14-tetramethylpentadecanedioic-7,7,8,9,9-d5 Acid
InChI formel InChI=1S/C19H36O5/c1-18(2,16(21)22)13-9-5-7-11-15(20)12-8-6-10-14-19(3,4)17(23)24/h15,20H,5-14H2,1-4H3,(H,21,22)(H,23,24)/i11D2,12D2,15D
SMIL CC(C)(CCCCC([2H])(C(O)([2H])C([2H])(CCCCC(C)(C(O)=O)C)[2H])[2H])C(O)=O
IUPAC navn 7,7,8,9,9-pentadeuterio-8-hydroxy-2,2,14,14-tetramethylpentadecanedioic acid
Molekylvægt (g/mol) 349.52
Formel vægt 349.29