Få mere at vide
Bemotrizinol, TRC
Artikelnummer. 30003903
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003903 100 mg
30003984 50 mg
30011258 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003903 Leverandør Toronto Research Chemicals Leverandørnr. TRCB131500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 187393-00-6
Molekylær formel C38 H49 N3 O5
Molekylvægt (g/mol) 627.81
Synonym 2,2'-[6-(4-Methoxyphenyl)-1,3,5-triazine-2,4-diyl]bis[5-[(2-ethylhexyl)oxy]phenol], 2,4-Bis[4-(2-ethylhexyloxy)-2-hydroxyphenyl]-6-(4-methoxyphenyl)-1,3,5-triazine, BEMT, Bemotrizinol, CGF 1607, FAT 70'884, Tinosorb S, Tinosorb S Aqua
IUPAC navn 5-(2-ethylhexoxy)-2-[4-[4-(2-ethylhexoxy)-2-hydroxyphenyl]-6-(4-methoxyphenyl)-1,3,5-triazin-2-yl]phenol
SMIL CCCCC(CC)COc1ccc(c(O)c1)c2nc(nc(n2)c3ccc(OCC(CC)CCCC)cc3O)c4ccc(OC)cc4
Tekniske data
Kemisk navn eller materiale Bemotrizinol
Analyt- eller komponentnavne Bemotrizinol
Renhedsgrad noter HPLC
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C38H49N3O5/c1-6-10-12-26(8-3)24-45-30-18-20-32(34(42)22-30)37-39-36(28-14-16-29(44-5)17-15-28)40-38(41-37)33-21-19-31(23-35(33)43)46-25-27(9-4)13-11-7-2/h14-23,26-27,42-43H,6-13,24-25H2,1-5H3
Løsningstype Neat
Industry Type Forensics
Mængde 100 mg
Formel vægt 627.3672
Procent renhed >95%