Få mere at vide
Belumosudil, MedChemExpress
Artikelnummer. 16479027
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
Pakningsstørrelse:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16479027 5 mg -
16466726 100 mg -
16442547 50 mg -
16406448 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16479027 Leverandør Medchem Express Leverandørnr. HY15307
MedChemExpress Belumosudil (KD025) is a selective inhibitor of ROCK2 with IC50s of 105 nM and 24 μM for ROCK2 and ROCK1, respectively. Anti-fibrotic properties.
Belumosudil (KD025) is a selective inhibitor of ROCK2 with IC50s of 105 nM and 24 μM for ROCK2 and ROCK1, respectively. Anti-fibrotic properties.
Tekniske data
Kemisk navn eller materiale Belumosudil
CAS 911417-87-3
Farve White
Renhedsgrad noter Research
Molekylær formel C26H24N6O2
Mængde 5 mg
Synonym KD025 SLx-2119
Opløselighedsinformation DMSO : 250 mg/mL (552.47 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMIL O=C(NC(C)C)COC1=CC=CC(C2=NC(NC3=CC4=C(NN=C4)C=C3)=C5C=CC=CC5=N2)=C1
Molekylvægt (g/mol) 452.51
Formel vægt 452.51
Procent renhed 98.23%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease