Få mere at vide
Bel-Art™ Spinbar™ Capsule Magnetic Stirring Bar
Artikelnummer. 11867473 Shop alle SP Scienceware produkter
Længde (metrisk):
5,3 cm
Pakningsstørrelse:
Hver
Artikelnummer. 11867473
Brand: Bel-Art™ F371520000
Capsule prevents stirring bar from spinning off center and damaging pH probes and gels.
  • Polygon magnetic stirring bar spins within polystyrene capsule
  • Prevents damage during pH adjustments and to gels during staining/destaining procedures
  • Large, flat capsule keeps stirring bar consistently centered on bottom of vertical electrophoresis units to efficiently mix the running buffer
  • Prevents stirring bar from being impeded by chemicals which, in water, transform into sticky gel and enables efficient dissolving
  • After use, capsule can be easily disassembled for cleaning
  • Autoclavable
Tekniske data
Autoklaverbar Autoclavable
Inkluderer 30 x 8mm polygon magnetic stirring bar
Belægning PTFE, FDA-grade, USP Class VI
Diameter (metrisk) 5.3 cm
Længde (metrisk) 5,3 cm
Materiale Polystyrene
Mængde 1
Form Polygon
Type Magnetic Stir Bar