Learn More
Bel-Art™ SP Scienceware™ Poxygrid™ Wire Test Tube Racks: 20-24mm
Chemical (organic solvents, salts, weak acids and weak alkalis) and corrosion-resistant steel wire racks are coated with heat-cured epoxy
Brand: Bel-Art™ F18767-0001
304.96 DKK valid until 2024-04-26
Use promo code "21699" to get your promotional price.
Additional Details : Weight : 0.24300kg
Description
- Withstands temperature range of -223°C (-369°F) to 121°C (250°F)
- Consists of square holes, three decks, a slotted grid bottom and extra strength reinforced edges
- Orange racks can be used for biohazard warning while yellow for radioactive warning
- Epoxy coatings are unaffected by organic solvents, salts, weak acids and weak alkalis
- Stores 40 tubes
- Autoclavable
Specifications
Tube Rack | |
Blue | |
Epoxy Coated Steel | |
10.43 x 4.57 x 3.25 in. (26.49 x 11.6 x 8.3cm) | |
10.35 in. | |
4.57 in. | |
3.25 in. |
Autoclavable | |
Tubes (20 to 24 mm) | |
40 | |
26.3 cm | |
11.6 cm | |
8.3 cm |